missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFSF10 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human TNFSF10 (NP_003801.1) (Val 114-Gly 281) fused to an initial Met

Brand:  Thermo Scientific™ 10409HNAE25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15295355

  • £124.92 / 25µg


Estimated Shipment
12-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Tumor Necrosis (Ligand) Receptor Superfamily Member 10 (TNFSF10), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 4-8ng/mL; 2. Measured in a cytotoxicity assay using NCI-H460 cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.2-0.8ng/mL
  • Formulation: Lyophilized in 40mM Tris 0.3M NaCl pH 7.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
19.6kDa
5 x 5μg
TNF (Tumor Necrosis Factor)
Tumor necrosis (ligand) receptor superfamily, member 10
Assay Standard
E. coli
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
APO2L, Apo-2L, CD253, TL2, TRAIL
TNFSF10
Cytokines & Receptors/CD Proteins
Untagged
Videos