missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ TNFRSF8 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human TNFRSF8 (NP_001234.2) extracellular domain (Met 1-Lys 379) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Invitrogen 10777H08H25

UNSPSC : 12352200

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15277165

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Tumor Necrosis Factor Receptor Superfamily Member 8 (TNFRSF8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFRSF8 at 2μg/mL (100μL/well) can bind human CD30L Fc Chimera with a linear range of 6.4-800ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
40kDa
5 x 5μg
TNF Receptors
TNFRSF8, CD30, D1S166E, Ki-1
TNFRSF8
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 8
Assay Standard
Mammalian