missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFRSF4 (CD134) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 216) of human TNFRSF4 (NP_003318.1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10481H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13415886

  • £236.31 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Tumor necrosis factor receptor superfamily, member 4 (TNFRSF4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF4-His at 2μg/mL (100μL/well) can bind human TNFSF4-mouse Fc, the ED50 of human TNFSF4-mouse Fc is 29ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
7293
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
TNF (Tumor Necrosis Factor)
TNFRSF4, RP5-902P8.3, ACT35, CD134, OX40, TXGP1L
TNFRSF4
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
21.7kDa
50μg
Truncated/Recombinant
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 4
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.