missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFRSF25 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human TNFRSF25 (NP_003781.1) extracellular domain (Met 1-Gln 199) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 12095H02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15285745

  • £200.92 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Tumor Necrosis Factor Receptor Superfamily, Member 25 (TNFRSF25), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8718
PAGE
50μg
C-Terminal
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 25
Assay Standard
Mammalian
Lyophilized
46kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
RP4-650H14.2, APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR
TNFRSF25
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.