missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFRSF21 Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Leu 350)of human DR6 (NP_055267.1) precursor fused to Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10175H02H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15254825

  • £159.28 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Tumor Necrosis Factor Receptor Superfamily, Member 21 (TNFRSF21), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; 2. Immobilized recombinant human DR6-Fc (Mfr. No. 10175-H02H) at 10μg/mL (100μL/well) can bind biotinylated human APP-Fc (Mfr. No. 10703-H02H) with a linear range of 0.03-0.25μg/mL
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27242
PAGE
50μg
C-Terminal
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 21
Assay Standard
Mammalian
Lyophilized
60.3kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
BM-018, DR6, mgC31965, TNFRSF21
TNFRSF21
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.