missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFRSF17 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TNFRSF17 (NP_001183.2) (Met 1-Ala 54) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus and a signal peptide at the N-terminus

Brand:  Thermo Scientific™ 10620H03H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15286995

  • £57.00 / 5µg


Estimated Shipment
08-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Tumor necrosis factor receptor superfamily, member 17 (TNFRSF17), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFSF13B/BAFF at 1μg/mL (100μL/well) can bind human TNFRSF17, the EC50 of human TNFRSF17 is 0.07μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
34kDa
5μg
TNF Receptors
TNFRSF17, BCM, BCMA, CD269
TNFRSF17
Cytokines & Receptors/CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 17
Assay Standard
Mammalian
Videos