missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFRSF10D (TRAILR4) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TNFRSF10D (NP_003831.2) (Met 1-His 211) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10413H08H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15285365

  • £494.48 / 250µg


Estimated Shipment
12-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Tumor necrosis factor receptor superfamily, member 10D, decoy with truncated death domain (TNFRSF10D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF10D at 10μg/mL (100μL/well) can bind biotinylated TNFSF10 with a linear range of 0.625-40ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
18.4kDa
5 x 50μg
TNF Receptors
TNFRSF10D, CD264, DCR2, TRAILR4, TRUNDD
TNFRSF10D
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain
Assay Standard
Mammalian
Videos