Sino Biological™ TNFRSF10B Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human TNFRSF10B (NP_003833.3) extracellular domain (Met 1-Glu 182) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10465H08H50

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15205475

Quantity Price
1 £ 153.0 / 50µg
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Common Name Tumor necrosis factor receptor superfamily, member 10b
Gene Symbol TNFRSF10B
Molecular Weight 15.8kDa
Quantity 50μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Tag His Tag
Gene Alias CD262, DR5, KILLER, KILLER/DR5, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9
Expression System Mammalian
Protein Subtype TNF Receptors
Endotoxin Concentration <1 EU/μg
Protein Family Cytokines & Receptors/CD Proteins
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Format Lyophilized

Tumor Necrosis Factor Receptor Superfamily Member 10B (TNFRSF10B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF10B at 10μg/mL (100μL/well) can bind biotinylated TNFSF10 with a linear range of 0.625-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.