missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFRSF10B Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human TNFRSF10B (NP_003833.3) extracellular domain (Met 1-Glu 182) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10465H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15275465

  • £123.88 / 25µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Tumor Necrosis Factor Receptor Superfamily Member 10B (TNFRSF10B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TNFRSF10B at 10μg/mL (100μL/well) can bind biotinylated TNFSF10 with a linear range of 0.625-20ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
15.8kDa
5 x 5μg
TNF Receptors
CD262, DR5, KILLER, KILLER/DR5, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9
TNFRSF10B
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 10b
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.