missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP2 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mature form of human TIMP2 (NP_003246.1) (Cys 27-Pro 220)
£309.00 - £462.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 22kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Endotoxin Concentration | <1 EU/μg |
Gene Alias | CSC-21K, TIMP2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15295325
|
Invitrogen
10396HNAH25 |
5 x 5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15225335
|
Invitrogen
10396HNAH50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
TIMP Metallopeptidase Inhibitor 2 (TIMP2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to inhibit human mMP-2 cleavage of a fluorogenic peptide substrate MCA-PLGL-DPA-AR-NH2 (R&D Systems Cat. No. ES001), the IC50 value is approximately 2.2nM
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
CSC-21K, TIMP2 | |
TIMP2 | |
Mammalian | |
Lyophilized | |
≥95% by SDS-PAGE |
22kDa | |
<1 EU/μg | |
TIMP metallopeptidase inhibitor 2 | |
Untagged | |
Protease Inhibitors | |
TIMP |
For Research Use Only. Not for use in diagnostic procedures.