missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ TGM3 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human TGM3 (Q08188) (Ala 2-Glu 693) fused to a polyhistidine tag at the N-terminus

Brand:  Invitrogen 11923H07B25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15255595

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Transglutaminase 3 (TGM3), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a synthetic peptide Benzyloxycarbonyl-Gln-Gly and NH2OH, the specific activity is >450pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, pH 8.5, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
78.8kDa
5 x 5μg
N-Terminal
<1 EU/μg
Transglutaminase 3
Assay Standard
His Tag
Frozen
≥90% by SDS-PAGE
Store at -80 °C. Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
MGC126249, mgC126250, TGE
TGM3
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.