missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TGM2 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human TGM2 (NP_004604.2) (Met 1-Ala 687) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11095H07B250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208235

  • £716.21 / 250µg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Transglutaminase 2 (C Polypeptide, Protein-Glutamine-Gamma-Glutamyltransferase) (TGM2), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 100mM NaCl, 2mM DTT, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
7052
PAGE
5 x 50μg
Full Length/Recombinant
RP5-1054A22.2, G-ALPHA-h, GNAH, TG2, TGC
TGM2
Other Proteins
His Tag
Frozen
79.6kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Transglutaminase 2 (C Polypeptide, Protein-Glutamine-Gamma-Glutamyltransferase)
Assay Standard
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.