Learn More
Thermo Scientific™ Sino Biological™ TFRC Recombinant Human Protein, N-His Tag
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TFRC (NP_003225.2) (Cys 89-Phe 760) fused to a polyhistidine tag at the N-terminus
Brand: Thermo Scientific™ 11020H07H50
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Transferrin Receptor (p90 CD71) (TFRC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized human TFRC at 10μg/mL (100μL/well) can bind human Transferrin, the EC50 of human Transferrin is 5.6ng/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
77.4kDa | |
50μg | |
Transferrin | |
CD71, TFR, TFR1, TRFR | |
TFRC | |
CD Proteins/Other Proteins | |
His Tag |
Lyophilized | |
≥90% by SDS-PAGE | |
Includes 5 vial of lyophilized protein. Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Transferrin receptor (p90, CD71) | |
Assay Standard | |
Mammalian |
For Research Use Only. Not for use in diagnostic procedures.