missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TFRC Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human TFRC (NP_003225.2) (Cys 89-Phe 760) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11020H07H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15207575

  • £787.00 / 250µg


Estimated Shipment
05-06-2018
Add to basket

Description

Description

Transferrin Receptor (p90 CD71) (TFRC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TFRC at 10μg/mL (100μL/well) can bind human Transferrin, the EC50 of human Transferrin is 5.6ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
77.4kDa
5 x 50μg
Transferrin
CD71, TFR, TFR1, TRFR
TFRC
CD Proteins/Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Includes 5 vial of lyophilized protein. Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Transferrin receptor (p90, CD71)
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.