missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TF (Transferrin) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human TF (NP_001054.1) (Met 1-Pro 698) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11019H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13483049

  • £275.87 / 50µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

Transferrin (TF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD71 at 10μg/mL (100μL/well) can bind human Transferrin, the EC50 of human Transferrin is 5.6ng/mL
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
76.6kDa
50μg
Full Length/Recombinant
<1 EU/μg
Transferrin
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Transferrin
TF, DKFZp781D0156, PRO1557, PRO2086
TF
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.