missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TACSTD2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 274) of human TROP2 (NP_002344.2) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10428H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15255395

  • £123.88 / 25µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Tumor-Associated Calcium Signal Transducer 2 (TACSTD2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4070
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Tumor-associated calcium signal transducer 2
Assay Standard
Mammalian
Lyophilized
29.4kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
TACSTD2, EGP-1, GA733, GA733-1, M1S1, TROP2
TACSTD2
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.