missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SUMO1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SUMO1 (AAH66306.1) (Ser 2-Val 101) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 13095H07E50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15206155

  • £419.53 / 50µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

SMT3 Suppressor of Mif Two 3 Homolog (SUMO1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
7341
PAGE
50μg
N-Terminal
<1 EU/μg
SMT3 suppressor of mif two 3 homolog
Assay Standard
E. coli
Lyophilized
12.4kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
OK/SW-cl.43, DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1
SUMO1
Other Proteins
His Tag
Videos