Learn More
SPINT1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human SPINT1 (O43278-2) (Met 1-Val 433) fused to a polyhistidine tag at the C-terminus
Brand: Invitrogen 11742H08H5
Additional Details : Weight : 0.10000kg
Description
Serine Peptidase Inhibitor Kazal Type 1 (SPINT1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (R&D Systems Cat. No. ES002); the IC50 value is <2nM
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5μg | |
<1 EU/μg | |
Serine peptidase inhibitor, Kazal type 1 | |
His Tag | |
Protease Inhibitors | |
Protease Inhibitors |
45.8kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
SPINT1, UNQ223/PRO256, HAI, HAI1, MANSC2 | |
SPINT1 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.