missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ SPARC Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SPARC (NP_003109.1) precursor (Met 1-Ile 303) fused to a carboxy-terminal polyhistidine tag

Brand:  Invitrogen 10929H08H50

UNSPSC : 12352202

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227405

  • £155.00 / 50µg


Estimated Shipment
14-08-2017
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Secreted Protein, Acidic, Cysteine-Rich Glycoprotein (SPARC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6678
PAGE
50μg
C-Terminal
<1 EU/μg
Secreted protein, acidic, cysteine-rich glycoprotein
Assay Standard
Mammalian
Lyophilized
34kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
ON, SPARC
SPARC
Other Proteins
His Tag