missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SOST Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human SOST (NP_079513.1) (Gln 24-Tyr 213) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10593H07H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15266965

  • £211.33 / 25µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Sclerostin (SOST), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50964
PAGE
5 x 5μg
N-Terminal
Wnt Proteins & Receptors
Sclerosteosis, SOST, VBCH
SOST
Other Proteins
His Tag
Lyophilized
22.5kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Sclerostin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.