missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SNAP25 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SNAP25 (P60880-1) (Met 1-Gly 206) fused to a polyhistidinetag at the N-terminus

Brand:  Thermo Scientific™ 11585H07E250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218695

  • £587.13 / 250µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Synaptosomal-Associated Protein 25kDa (SNAP25), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6616
PAGE
5 x 50μg
N-Terminal
<1 EU/μg
Synaptosomal-associated protein
Assay Standard
E. coli
Lyophilized
24.8kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
FLJ23079, RIC-4, RIC4, SEC9, SNAP, SNAP-25, bA416N4.2, dJ1068F16.2
SNAP25
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.