missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SLPI Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the secreted form of human SLPI (P03973) (Ser 26-Ala 132) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10343H07B5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15205245

  • £47.52 / 5µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Secretary Leukocyte Peptidase Inhibitor (SLPI), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6590
PAGE
5μg
Full Length/Recombinant
ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4
SLPI
Proteases, Peptidases & Inhibitors
His Tag
Frozen
14kDa
>84% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Secretory Leukocyte Peptidase Inhibitor
Assay Standard
Insect
Videos