Learn More
Thermo Scientific™ Sino Biological™ SLAMF7 Recombinant Human Protein, His Tag
This recombinant protein was expressed from a DNA sequence encoding the human SLAMF7 (NP_067004.3) extracellular domain (Met 1-Met 226) fused to a polyhistidine tag at the C-terminus
Brand: Thermo Scientific™ 11691H08H50
Additional Details : Weight : 0.10000kg
Description
Slam Family Member 7 (SLAMF7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to bind biotinylated human SLAMF7 in a functional ELISA
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories | |
23.8kDa | |
50μg | |
Other Proteins | |
UNQ576/PRO1138, 19A, CD319, CRACC, CS1 | |
SLAMF7 | |
CD Proteins | |
His Tag |
Lyophilized | |
≥90% by SDS-PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
SLAM family member 7 | |
Assay Standard | |
Mammalian |
For Research Use Only. Not for use in diagnostic procedures.