missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAMF1 (CD150) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human SLAMF1 precursor (NP_003028.1), residues 1 to 258, fused to a polyhistidine tag at the C-terminus
£58.00 - £874.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Gene ID (Entrez) | 6504 |
Molecular Weight (g/mol) | 25.8kDa |
Storage Requirements | Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Structural Form | Truncated/Recombinant/Cell Surface Protein/Ligand |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13405986
|
Invitrogen
10837H08H5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13495976
|
Invitrogen
10837H08H250 |
5 x 50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Signaling lymphocytic activation molecule (SLAMF1/CD150), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.- Activity: Measured by its binding ability in a functional ELISA; immobilized human SLAMF1-His at 1μg/well can bind biotinylated recombinant human SH2D1A with a linear range of 0.8-20.0μg/mL
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
25.8kDa | |
Truncated/Recombinant/Cell Surface Protein/Ligand | |
CD150, CDw150, SLAM, SLAMF1 | |
SLAMF1 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
6504 | |
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
<1 EU/μg | |
Signaling lymphocytic activation molecule family member 1 | |
His Tag | |
CD Proteins | |
Glycoproteins |
For Research Use Only. Not for use in diagnostic procedures.