missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SIGIRR Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse SIGIRR (NP_075546.2) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50122M03H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15322756

  • £123.88 / 25µg


Estimated Shipment
10-07-2018
Add to basket

Includes: Five vials of protein in suspension buffer

Description

Description

Single Immunoglobulin and Toll-Interleukin 1 Receptor (TIR) Domain (SIGIRR), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
24058
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Single immunoglobulin and toll-interleukin 1 receptor (TIR) domain
Assay Standard
Mammalian
Lyophilized
40.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
AI256711, mgC102426, TIR8
SIGIRR
Cytokines & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.