Sino Biological™ SFTPD Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse SFTPD (NP_033186.1) precursor (Met 1-Phe 374) fused to a C-terminal polyhistidine tag

Overview
Brand: Invitrogen

Manufacturer Part Number: 50205M08H50

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: One vial of protein in suspension buffer

Product Code. 15352936

Quantity Price
1 £ 212.0 / 50µg
Estimated Shipment date
from Supplier 07-04-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purification Method PAGE
Protein Tag His Tag
Protein Family Cell Structure/Organelle Proteins
Molecular Weight 37.2kDa
Common Name Surfactant, pulmonary-associated protein D
Format Lyophilized
Structural Form Full Length/Recombinant
Gene Symbol SFTPD
Quantity 50μg
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Expression System Mammalian
Gene ID (Entrez) 20390
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias AI573415, SP-D, Sftp4, Sftpd
Termination C-Terminal
Endotoxin Concentration <1 EU/μg

Surfactant, Pulmonary-Associated Protein D (SFTPD), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible