Sino Biological™ SFTPD Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SFTPD (NP_003010.4) precursor (Met 1-Phe 375) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11041H08H50

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15207625

Quantity Price
1 £ 198.0 / 50µg
Estimated Shipment date
from Supplier 12-04-2017
Add to basket


Description and Specification

Specification

Termination C-Terminal
Quantity 50μg
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Common Name Surfactant, pulmonary-associated protein D
Expression System Mammalian
Protein Tag His Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purification Method PAGE
Molecular Weight 37kDa
Protein Family Cell Structure/Organelle Proteins
Purity ≥85% by SDS-PAGE
Format Lyophilized
Structural Form Full Length/Recombinant
Gene Alias COLEC7, PSP-D, SFTP4, SP-D
Gene ID (Entrez) 6441
Gene Symbol SFTPD

Surfactant, Pulmonary-Associated Protein D (SFTPD), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.