missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SERPINE1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SerpinE1 precursor (NP_000593.1) (Met 1-Pro 402) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10296H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15235115

  • £469.50 / 50µg


EU Stock 3
Add to basket

Description

Description

Serpin Peptidase Inhibitor, Clade E, Member 1 (SERPINE1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM NaAc, 100mM NaCl, pH5.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5054
PAGE
50μg
C-Terminal
<1 EU/μg
Serpin peptidase inhibitor, clade E, member 1
Assay Standard
Mammalian
Lyophilized
44.2kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
SERPINE1, PAI, PAI-1, PAI1, PLANH1
SERPINE1
Proteases, Peptidases & Inhibitors
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.