Sino Biological™ SERPINE1 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SerpinE1 precursor (NP_000593.1) (Met 1-Pro 402) fused to a C-terminal polyhistidine tag

Overview
Brand: Invitrogen

Manufacturer Part Number: 10296H08H250

RECOM HU SERPINE1 (HIS)

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15215115

Quantity Price
1 £ 1500.0 / 250µg
Estimated Shipment date
from Supplier 08-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 44.2kDa
Purity ≥95% by SDS-PAGE
Structural Form Full Length/Recombinant
Gene Symbol SERPINE1
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Purification Method PAGE
Quantity 5 x 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Endotoxin Concentration <1 EU/μg
Gene Alias SERPINE1, PAI, PAI-1, PAI1, PLANH1
Common Name Serpin peptidase inhibitor, clade E, member 1
Protein Tag His Tag
Expression System Mammalian
Format Lyophilized
Gene ID (Entrez) 5054

Serpin Peptidase Inhibitor, Clade E, Member 1 (SERPINE1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM NaAc, 100mM NaCl, pH5.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.