Sino Biological™ SERPINB8 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse SERPINB8 (NP_035589.1) (Met 1-Pro 374) fused to a C-terminal polyhistidine tag

Overview
Brand: Invitrogen

Manufacturer Part Number: 50215M08B5

5UG RECOM MS SERPINB8 (HIS)

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.
Includes: One vial of protein in suspension buffer

Product Code. 15302956

Quantity Price
1 £ 62.5 / 5µg
Estimated Shipment date
from Supplier 03-02-2017
Add to basket


Description and Specification

Specification

Molecular Weight 43.6kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Termination C-Terminal
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias Serpinb8, CAP-2, CAP2, NK10, Spi8, ovalbumin
Common Name Serine peptidase inhibitor, clade B, member 8
Gene Symbol SERPINB8
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 20725

Serine Peptidase Inhibitor, Clade B, Member 8 (SERPINB8), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.