missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SERPINA3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SERPINA3 (NP_001076.2) (Met 1-Ala 423) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10307H08H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15285135

  • £138.46 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Serine Peptidase Inhibitor Clade A Member 3 (SERPINA3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Anaspec Cat. No. 27114), the IC50 value is <20nM
  • Formulation: Lyophilized in 25mM HEPES, 0.15M NaCl, pH 7.8, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
46.5kDa
5μg
Proteinase Inhibitor
SERPINA3, AACT, ACT, GIG24, GIG25, mgC88254
SERPINA3
Protease Inhibitors
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Serine peptidase inhibitor, clade A, member 3
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.