missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SEMA5A Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Met 1-Thr 765) of human SEMA5A (NP_003957.2) extracellular domain fused to Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 11300H02H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208505

  • £211.33 / 25µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Sema Domain, Seven Thrombospondin Repeats (TM) and Short Cytoplasmic Domain, (Semaphorin) 5A (SEMA5A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, 100mMM EDTA, 0.05mM Tween20, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
9037
PAGE
5 x 5μg
C-Terminal
Other Proteins
FLJ12815, SEMAF, semF, SEMA5A
SEMA5A
Other Proteins
IgG1 Fc Tag
Lyophilized
110.7kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Sema domain, 7 thrombospondin repeats, TM and short cytoplasmic domain, (semaphorin) 5A
Assay Standard
Mammalian
Videos