missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SEMA4A Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human SEMA4A (NP_071762.2) (Met 1-His 683) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11756H08H250

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15219125

  • £693.31 / 250µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

Sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4 (SEMA4A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated mouse SEMA4D in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
73.4kDa
5 x 50μg
Other Proteins
RP11-54H19.2, CORD10, FLJ12287, RP35, SEMAB, SEMB
SEMA4A
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Sema domain, immunoglobulin domain (Ig), TM and short cytoplasmic domain, (semaphorin) 4A
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.