missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SELL Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 332) of mouse SELL (NP_035476.1) precursor fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50045M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15342586

  • £57.26 / 5µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Selectin L (SELL), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
20343
PAGE
5μg
C-Terminal
Other Proteins
Sell, AI528707, CD62L, L-selectin, LECAM-1, Lnhr, Ly-22, Ly-m22, Lyam-1, Lyam1
SELL
Other Proteins
His Tag
Lyophilized
34.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
<1 EU/μg
Selectin L
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.