missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SELL (CD62L) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 332) of human SELL (P14151-1) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 11838H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13485596

  • £187.38 / 50µg


Estimated Shipment
10-05-2018
Add to basket

Description

Description

L-Selectin (SELL/CD62L), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human SELL-His at 2μg/well can bind biotinylated human CD34-His with a linear range of 0.016-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6402
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Lectins & Leptins
RP1-117P20.1, CD62L, LAM1, LECAM1, LEU8, LNHR, LSEL, LYAM1, PLNHR, TQ1
SELL
CD Proteins
His Tag
Lyophilized
34.5kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Selectin L
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.