missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SELE (E-SELECTIN) Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 556) of the human SELE precursor (NP_000441.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10335H03H5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15205205

  • £56.74 / 5µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Selectin E (SELE) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.&line-break;Background: E-selectin, also known as endothelial leukocyte adhesion molecule-1 (ELAM-1) and CD62E, is an inducible adhesion molecule that is expressed at relatively high density on the surface of cultured endothelial cells that have been activated by proinflammatory cytokines, such as IL- 1β, TNF-α, or bacterial endotoxin. E-selectin exhibits a complex mosaic structure consisting of a large extracellular region comprised of a lectin domain, an EGF-like domain, and a short consensus repeat (SCR) domain, followed by a transmembrane region and a relatively short (32 aa) cytoplasmic tail. As a member of the LEC-CAM or selectin family, E-selectin recognises and binds to sialylated carbohydrates including members of the Lewis X and Lewis A families found on monocytes, granulocytes, and T-lymphocytes. E-selectin supports rolling and stable arrest of leukocytes on activated vascular endothelium, and furthermore, it was indicated that it can also transduce an activating stimulus via the MAPK cascade into the endothelial cell during leukocyte adhesion. In addition, this adhesion molecule appears to be involved in the pathogenesis of atherosclerosis.
  • N-terminal Sequence Analysis: Trp 22
  • Activity: measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when cells are added to SELE coated plates (2μg/mL, 100μL/well) approximately 90-100 % will adhere specifically
  • Formulation: lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
86.5kDa
5μg
Other Proteins
SELE, RP1-117P20.2, CD62E, ELAM, ELAM1, ESEL, LECAM2
SELE
Other Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
SELE selectin E
Assay Standard
Mammalian
Videos