missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SDC1 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse SDC1 (NP_035649.1) (Met 1-Glu 252) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50641M08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15339086

  • £203.00 / 50µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of protein in suspension buffer

Description

Description

Syndecan 1 (SDC1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
20969
PAGE
50μg
C-Terminal
<1 EU/μg
Syndecan 1
Assay Standard
Mammalian
Lyophilized
26kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
SDC1, AA408134, AA409076, CD138, Sstn, Synd, Synd1, syn-1
SDC1
CD Proteins
His Tag
Videos