missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SCARB2 Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse SCARB2 (NP_031670.1) (Met 1-Thr 432) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50052M08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15312606

  • £718.29 / 250µg


Estimated Shipment
08-06-2018
Add to basket

Includes: Five vials of protein in suspension buffer

Description

Description

Scavenger Receptor Class B, Member 2 (SCARB2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris-Citrate, 300mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12492
PAGE
5 x 50μg
C-Terminal
<1 EU/μg
Scavenger receptor class B, member 2
Assay Standard
Mammalian
Lyophilized
47.8kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
Scarb2, 9330185J12Rik, Cd36l2, LGP85, LIMP-2, MLGP85
SCARB2
CD Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.