Sino Biological™ SCARB2 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SCARB2 (NP_005497.1) extracellular domain (Arg 27-Thr 432) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11063H03H50

SCARB2 RECOMBINANT HUMAN PROTEIN,HIGG1-FC.HIS TAG

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15217695

Quantity Price
1 £ 188.0 / 50µg
Estimated Shipment date
from Supplier 09-03-2017
Add to basket


Description and Specification

Specification

Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias SCARB2, AMRF, CD36L2, HLGP85, LIMPII, SR-BII
Common Name Scavenger receptor class B, member 2
Gene Symbol SCARB2
Protein Tag His Tag/IgG1 Fc Tag
Protein Family CD Proteins
Molecular Weight 74.4kDa
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Scavenger receptor class B, member 2 (SCARB2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant human RSPO1 in a functional ELISA
  • Formulation: Lyophilized in 50mM Tris-Citrate, 300mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.