missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ SCARB2 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the human SCARB2 (NP_005497.1) extracellular domain (Arg 27-Thr 432) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 11063H03H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15287685

  • £682.90 / 250µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Scavenger receptor class B, member 2 (SCARB2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant human RSPO1 in a functional ELISA
  • Formulation: Lyophilized in 50mM Tris-Citrate, 300mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
74.4kDa
5 x 50μg
Other Proteins
SCARB2, AMRF, CD36L2, HLGP85, LIMPII, SR-BII
SCARB2
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Scavenger receptor class B, member 2
Assay Standard
Mammalian
Videos