Learn More
S100B Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to a polyhistidine tag at the N-terminus
Brand: Invitrogen 10181H07E250
Code : NEW
Additional Details : Weight : 0.10000kg
Description
S100 Calcium Binding Protein B (S100B), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: 1. Measured by its ability to bind human AGER in a functional ELISA; 2. Measured by its ability to bind TP53 in a functional ELISA
- Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.5
- Reconstitution: No reconstitution needed
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5 x 50μg | |
NEF, S100, S100beta | |
S100B | |
E. coli | |
Neurotrophic Factors, Other Proteins |
12kDa | |
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
S100 calcium binding protein B | |
His Tag | |
Frozen | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.