missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100B Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10181H07E25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15254855

  • £122.84 / 25µg


Estimated Shipment
13-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

S100 Calcium Binding Protein B (S100B), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind human AGER in a functional ELISA; 2. Measured by its ability to bind TP53 in a functional ELISA
  • Formulation: PBS, 0.2μg/μL BSA, 10% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12kDa
5 x 5μg
Neurotrophic Factors|Other Proteins
S100 calcium binding protein B
Assay Standard
His Tag
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
NEF, S100, S100beta
S100B
E. coli
Videos