missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100B Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human S100B (NP_006263.1) (Ser 2-Glu 92) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10181H01H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15224855

  • £127.01 / 25µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

S100 calcium binding protein B (S100B), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a functional ELISA: 1. Measured by its ability to bind mouse S100A1; 2. Measured by its ability to bind TP53
  • Formulation: Lyophilized in 100mM Gly, 10mM NaCl, 50mM Tris, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
37.2kDa
5 x 5μg
Neurotrophic Factors|Other Proteins
NEF, S100, S100beta
S100B
Other Proteins
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
S100 calcium binding protein B
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.