missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A9 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human S100A9 (NP_002956.1) (Met 1-Pro 114)

Brand:  Thermo Scientific™ 11145HNAE5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15238305

  • £73.14 / 5µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

S100 Calcium Binding Protein A9 (S100A9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells; the ED50 for this effect is typically 3-15μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
26kDa
5μg
Other Proteins
S100 calcium binding protein A9
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14
S100A9
Other Proteins
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.