missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A8 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human S100A8 (NP_002955.2) (Leu 2-Glu 93) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11138H07E250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15228275

  • £647.51 / 250µg

Estimated Shipment
Add to basket



S100 Calcium Binding Protein A8 (S100A8), recombinant human protein is supplied in frozen format. It is suitable for use in performing cell-based assays and protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind recombinant human S100A9 in a functional ELISA; measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells, the ED50 for this effect is typically 10-30μg/mL
  • Formulation: 20mM Tris, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 50μg
Other Proteins
S100 calcium binding protein A8
Assay Standard
His Tag
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
E. coli

For Research Use Only. Not for use in diagnostic procedures.