missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A6 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human S100A6 (NP_055439.1) (Met 1-Gly 90) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10939H08B25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15207425

  • £204.04 / 25µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

S100 Calcium Binding Protein A6 (S100A6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, 0.5mM PMSF, 1mM TCEP, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6277
PAGE
5 x 5μg
C-Terminal
Other Proteins
2A9, 5B10, CABP, CACY, PRA, S100A6
S100A6
Other Proteins
His Tag
Lyophilized
12kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
<1 EU/μg
S100 calcium binding protein A6
Assay Standard
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.