missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A4 Recombinant Human Protein, N-hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the human S100A4 (NP_002952.1) (Met 1-Lys 101) fused to Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10185H01H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15214865

  • £57.26 / 5µg


EU Stock 1
Add to basket

Description

Description

S100 Calcium Binding Protein A4 (S100A4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6275
PAGE
5μg
N-Terminal
Other Proteins
18A2, 42A, CAPL, FSP1, MTS1, P9KA, PEL98
S100A4
Other Proteins
IgG1 Fc Tag
Lyophilized
38.4kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
S100 calcium binding protein A4
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.