missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A13 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the native human S100A13 (NP_001019381.1) (Met 1-Lys 98)

Brand:  Thermo Scientific™ 10991HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15257495

  • £206.12 / 50µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

S100 Calcium Binding Protein A13 (S100A13), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
6284
PAGE
50μg
Full Length/Recombinant
OTTHUMP00000034802, S100 calcium-binding protein A13, protein S100-A13
S100A13
Other Proteins
Lyophilized
11.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
S100 calcium binding protein A13
Assay Standard
E. coli
Videos