Learn More
S100A12 Recombinant Human Protein, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the native human S100A12 (NP_005612.1) (Met 1-Glu 92)
Brand: Invitrogen 11143HNAE50
Code : NEW
Additional Details : Weight : 0.10000kg
Description
S100 Calcium Binding Protein A12 (S100A12), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human S100A12 at 2μg/mL (100μL/well) can bind human AGER with a linear range of 0.032-20μg/mL
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
50μg | |
S100A12, CAAF1, CAGC, CGRP, ENRAGE, MRP6, p6 | |
S100A12 | |
E. coli | |
Lyophilized | |
≥90% by SDS-PAGE |
10.6kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
S100 calcium binding protein A12 | |
Untagged | |
Other Proteins | |
Other Proteins |
For Research Use Only. Not for use in diagnostic procedures.