missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ S100A12 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the native human S100A12 (NP_005612.1) (Met 1-Glu 92)

Brand:  Thermo Scientific™ 11143HNAE5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15218295

  • £56.21 / 5µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

S100 Calcium Binding Protein A12 (S100A12), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human S100A12 at 2μg/mL (100μL/well) can bind human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
10.6kDa
5μg
Other Proteins
S100 calcium binding protein A12
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
S100A12, CAAF1, CAGC, CGRP, ENRAGE, MRP6, p6
S100A12
Other Proteins
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.